missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLC37A4 Polyclonal specifically detects SLC37A4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | SLC37A4 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?