missing translation for 'onlineSavingsMsg'
Learn More

Smad2, p Ser467 Antibody - BSA Free, R&D Systems™

Product Code. 18666758 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
Unit Size:
0.03mg
0.05mg
0.1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18666758 0.025 mg 0.03mg
18292954 0.05 mg 0.05mg
18220591 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18666758 Fournisseur Novus Biologicals Code fournisseur NBP2547710.025mg
 Restricted Item
Estimated Shipment: 02-12-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Smad2 Polyclonal specifically detects Smad2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Simple Western, Western Blot.
TRUSTED_SUSTAINABILITY

Spécification

Antigen Smad2
Applications Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl and 50% glycerol with 0.02% Sodium Azide
Gene Alias hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, pSMAD, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2
Gene Symbols SMAD2
Host Species Rabbit
Immunogen Smad2 (Phospho-Ser467) antibody was raised against a peptide sequence around phosphorylation site of serine 467 (C-S-S-M-S (p) ) derived from Human Smad2.
Molecular Weight of Antigen 60 kDa
Purification Method Epitope affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Metastasis, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4087
Test Specificity This antibody detects endogenous level of Smad2 only when phosphorylated at serine 467.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.