missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NB100-56440SS
This item is not returnable.
View return policy
Description
SMAD6 Polyclonal specifically detects SMAD6 in Human, Mouse, Rat, Primate, Monkey, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SMAD6 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 4091 | |
| Human, Mouse, Rat, Primate, Monkey, Sheep | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 0.5 - 2 ug/mL. Use reported by customer review, Simple Western 1:50, Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1 - 5 ug/mL. Use reported in scientific literature (PMID 20032053), Immunohistochemistry-Paraffin 5 - 10 ug/mL. Use reported in scientific literature (PMID 20032053) | |
| O43541 | |
| SMAD6 | |
| This antibody was developed against 2 synthetic peptides found between amino acids 30-150 and 300-400 of human SMAD6 (GenBank Accession No. AAH12986.1). These peptide sequences are specific to SMAD6, and have high homology to SMAD6 in multiple species. | |
| 0.025 mg | |
| Primary | |
| A 52 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction