missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
188.00€ - 526.00€
Specifications
| Antigen | SMAD6 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5 - 2 ug/mL. Use reported by customer review, Simple Western 1:50, Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1 - 5 ug/mL. Use reported in scientific literature (PMID 20032053), Immunohistochemistry-Paraffin 5 - 10 ug/mL. Use reported in scientific literature (PMID 20032053) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18706563
|
Novus Biologicals
NB100-56440SS |
0.025 mg |
188.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799393
|
Novus Biologicals
NB100-56440 |
0.1 mg |
526.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMAD6 Polyclonal specifically detects SMAD6 in Human, Mouse, Rat, Primate, Monkey, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SMAD6 | |
| Western Blot 0.5 - 2 ug/mL. Use reported by customer review, Simple Western 1:50, Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1 - 5 ug/mL. Use reported in scientific literature (PMID 20032053), Immunohistochemistry-Paraffin 5 - 10 ug/mL. Use reported in scientific literature (PMID 20032053) | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6 | |
| SMAD6 | |
| IgG | |
| Protein G purified | |
| A 52 kDa band is observed. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Primate, Monkey, Sheep | |
| O43541 | |
| 4091 | |
| This antibody was developed against 2 synthetic peptides found between amino acids 30-150 and 300-400 of human SMAD6 (GenBank Accession No. AAH12986.1). These peptide sequences are specific to SMAD6, and have high homology to SMAD6 in multiple species. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title