Learn More
SMAD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand:  Novus Biologicals NB100-56440
 View more versions of this product
Product Code. 18799393
  • 526.00€ / 0.10mg
Description
SMAD6 Polyclonal specifically detects SMAD6 in Human, Mouse, Rat, Primate, Monkey, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot 0.5 - 2 ug/mL. Use reported by customer review, Simple Western 1:50, Immunohistochemistry 5 - 10 ug/mL, Immunocytochemistry/Immunofluorescence 1 - 5 ug/mL. Use reported in scientific literature (PMID 20032053), Immunohistochemistry-Paraffin 5 - 10 ug/mL. Use reported in scientific literature (PMID 20032053)
SMAD6
0.1 mg
Rat, Primate
Polyclonal
hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6
Protein G purified