missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SMCX Antibody (PCRP-KDM5C-1A11), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26752-20ug

 View more versions of this product

Product Code. 30043392

  • 232.00€ / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SMCX Monoclonal antibody specifically detects SMCX in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SMCX
Monoclonal
0.2 mg/mL
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
8242
Store at 4°C.
IgG2c
Flow Cytometry
PCRP-KDM5C-1A11
Unconjugated
DXS1272EMRXJ, EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1C, JARID1Clysine-specific demethylase 5C, jumonji, AT rich interactive domain 1C, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, lysine (K)-specific demethylase 5C, MRXSJ, Protein SmcX, Protein Xe169, Smcx homolog, X chromosome, SMCXselected cDNA on X, Smcy homolog, X-linked, Smcy homolog, X-linked (mouse), XE169JmjC domain-containing protein SMCX
Recombinant full-length human SMCX protein
20 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.