missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SMCX Antibody (PCRP-KDM5C-1A11), Novus Biologicals™
Mouse Monoclonal Antibody
232.00€ - 462.00€
Specifications
| Antigen | SMCX |
|---|---|
| Clone | PCRP-KDM5C-1A11 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043392
|
Novus Biologicals™
NBP3-26752-20ug |
20 μg |
232.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043750
|
Novus Biologicals™
NBP3-26752-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMCX Monoclonal antibody specifically detects SMCX in Human samples. It is validated for Flow CytometrySpecifications
| SMCX | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 8242 | |
| IgG2c | |
| Protein A or G purified |
| PCRP-KDM5C-1A11 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| DXS1272EMRXJ, EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1C, JARID1Clysine-specific demethylase 5C, jumonji, AT rich interactive domain 1C, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, lysine (K)-specific demethylase 5C, MRXSJ, Protein SmcX, Protein Xe169, Smcx homolog, X chromosome, SMCXselected cDNA on X, Smcy homolog, X-linked, Smcy homolog, X-linked (mouse), XE169JmjC domain-containing protein SMCX | |
| Recombinant full-length human SMCX protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title