missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAPC4 Antibody (PCRP-SNAPC4-3A7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23950-20ug
This item is not returnable.
View return policy
Description
SNAPC4 Monoclonal antibody specifically detects SNAPC4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| SNAPC4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry : 1-2 μg/million cells, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| FLJ13451, Proximal sequence element-binding transcription factor subunit alpha, PSE-binding factor subunit alpha, PTF subunit alpha, PTFalpha, small nuclear RNA activating complex, polypeptide 4, 190kD, small nuclear RNA activating complex, polypeptide 4, 190kDa, SNAP190SNAPc 190 kDa subunit, SNAPc subunit 4, snRNA-activating protein complex 190 kDa subunit, snRNA-activating protein complex subunit 4 | |
| Recombinant full-length human SNAPC4 protein | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-SNAPC4-3A7 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6621 | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction