missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAPC4 Antibody (PCRP-SNAPC4-3A7), Novus Biologicals™
Mouse Monoclonal Antibody
232.00€ - 462.00€
Specifications
| Antigen | SNAPC4 |
|---|---|
| Clone | PCRP-SNAPC4-3A7 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry : 1-2 μg/million cells, Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621024
|
Novus Biologicals
NBP3-23950-20ug |
20 μg |
232.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630214
|
Novus Biologicals
NBP3-23950-100ug |
100 μg |
462.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNAPC4 Monoclonal antibody specifically detects SNAPC4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
| SNAPC4 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ13451, Proximal sequence element-binding transcription factor subunit alpha, PSE-binding factor subunit alpha, PTF subunit alpha, PTFalpha, small nuclear RNA activating complex, polypeptide 4, 190kD, small nuclear RNA activating complex, polypeptide 4, 190kDa, SNAP190SNAPc 190 kDa subunit, SNAPc subunit 4, snRNA-activating protein complex 190 kDa subunit, snRNA-activating protein complex subunit 4 | |
| Recombinant full-length human SNAPC4 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-SNAPC4-3A7 | |
| Flow Cytometry : 1-2 μg/million cells, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6621 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title