missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNIP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15785751 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15785751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15785751 Supplier Invitrogen™ Supplier No. PA557864

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QHREPSEQEH RRARNSDRDR HRGHSHQRRT SNERPGSGQG QGRDRDTQNL QAQEEEREFY NARRREHRQR NDVGGG Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNIP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNIP1
Gene Accession No. Q8TAD8
Gene Alias 2410133M08Rik; dJ423B22.2; FHA domain-containing protein SNIP1; PMRED; RP3-423B22.3; Smad nuclear interacting protein; smad nuclear interacting protein 1; Smad nuclear interacting protein 1 Snip1; smad nuclear-interacting protein 1; Snip1
Gene Symbols SNIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNIP1. Recombinant protein control fragment (Product #RP-94439).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79753
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.