missing translation for 'onlineSavingsMsg'
Learn More

SNX16 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Codice prodotto. 18559946 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantity:
0.1 mL
Dimensione della confezione:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18559946 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18559946 Fornitore Novus Biologicals N. del fornitore NBP318461AF488

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

SNX16 Polyclonal antibody specifically detects SNX16 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifica

Antigen SNX16
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp666H147, sorting nexin 16, sorting nexin-16
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 250 and 300..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64089
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.