missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX16 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Description
SNX16 Polyclonal antibody specifically detects SNX16 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | SNX16 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 585 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp666H147, sorting nexin 16, sorting nexin-16 |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between Between 250 and 300.. |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?