missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNX20 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15778281 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15778281 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15778281 Supplier Invitrogen™ Supplier No. PA560451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PGCMGPITQC TARTQQEAPA TGPDLLHPGP DGHLDTHSGL SSNSSMTTRE LQQYWQNQKC RWKHVKLLFE IASAR Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 60%.

SNX20 interacts with the cytoplasmic domain of PSGL1 and cycles PSGL1 into endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX20
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX20
Gene Accession No. Q7Z614
Gene Alias 9130017C17Rik; RGD1307787; selectin ligand interactor cytoplasmic 1; selectin ligand interactor cytoplasmic-1; selectin ligand-interactor cytoplasmic 1; SLIC1; SLIC-1; SNX20; sorting nexin 20; sorting nexin-20
Gene Symbols SNX20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNX20. Recombinant protein control fragment (Product #RP-98742).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124460
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.