Learn More
Sodium Bromide, TRC
Product Code. 30008083
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008083 10 g 10g
30003281 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008083 Supplier Toronto Research Chemicals Supplier No. TRCS61516010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7647-15-6
Molecular Formula Br Na
Molecular Weight (g/mol) 102.89
Synonym Sodium bromide, NSC 77384, Sanibrom 40, Sedoneural
IUPAC Name bromosodium
SMILES [Na]Br
Specifications
Quantity 10 g
InChI Formula InChI=1S/BrH.Na/h1H;/q;+1/p-1
Formula Weight 101.9081
Chemical Name or Material Sodium Bromide