missing translation for 'onlineSavingsMsg'
Learn More

SOWAHD Antibody, DyLight 680, Novus Biologicals™

Product Code. 18559343 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18559343 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18559343 Supplier Novus Biologicals Supplier No. NBP298173FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOWAHD Polyclonal antibody specifically detects SOWAHD in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOWAHD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias ANKRD58, Ankyrin Repeat Domain 58, Ankyrin Repeat Domain-Containing Protein 58, Ankyrin Repeat Domain-Containing Protein SOWAHD, Protein Sosondowah Homolog D, Sosondowah Ankyrin Repeat Domain Family Member D
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SOWAHD fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347454
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.