missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SOX2 Monoclonal specifically detects SOX2 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | SOX2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 017 |
| Conjugate | Biotin |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the human SOX2. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?