missing translation for 'onlineSavingsMsg'
Learn More

SOX2 Antibody (PO00-28), Novus Biologicals™

Product Code. 30115488 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30115488 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30115488 Supplier Novus Biologicals Supplier No. NBP332972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SOX2 Monoclonal antibody specifically detects SOX2 in Human,Mouse,Rat samples. It is validated for ChIP assay,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX2
Applications ChIP Assay, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone PO00-28
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Chromatin Immunoprecipitation Use 0.5-2 μg for 25 μg of chromatin, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250, Immunohistochemistry-Paraffin 1:1000-1:4000
Formulation PBS (pH7.4), 0.1% BSA and 40% Glycerol
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Host Species Rabbit
Immunogen Recombinant protein within human SOX2 aa 1-317. (Uniprot: P48431)
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6657
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.