missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sp56 Monoclonal Antibody (4B2)
GREENER_CHOICE

Product Code. 11583453
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11583453 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11583453 Supplier Invitrogen™ Supplier No. MA110868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10868 detects mouse Sperm Protein sp56 from mouse samples. This antibody does not cross react with human sperm. MA1-10868 has been successfully used in ELISA, Western blot, and immunohistochemistry applications. This antibody will detect denatured, non-reduced sp56, but will not detect the denatured, reduced protein. The MA1-10868 immunogen is a mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sp56
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4B2
Concentration ∼1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Zp3r
Gene Accession No. Q60736
Gene Alias AC122773.1; AV264399; SP56; sperm fertilization protein 56; zona pellucida 3 receptor; zona pellucida sperm-binding protein 3 receptor; Zp3r
Gene Symbols Zp3r
Host Species Mouse
Immunogen Mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22789
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.