Learn More
Spermidine synthase Mouse, anti-Human, Clone: 2F4E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68548-1-IG-150UL
Description
The Spermidine synthase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Spermidine synthase. This antibody recognizes human antigen. The Spermidine synthase antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| Spermidine synthase | |
| 2F4E10 | |
| PAPT, SPDSY, spermidine synthase, SPS1, SRM, SRML1 | |
| 150 μL | |
| 6723 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2b |