Learn More
Spermidine synthase Mouse, anti-Human, Clone: 2F4E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68548-1-IG-150UL
 View more versions of this product
Product Code. 18075145
  • 410.00€ / 150µL
Description
The Spermidine synthase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Spermidine synthase. This antibody recognizes human antigen. The Spermidine synthase antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
Spermidine synthase
2F4E10
PAPT, SPDSY, spermidine synthase, SPS1, SRM, SRML1
150 μL
6723
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunofluorescence, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa
IgG2b