Learn More
Spermidine synthase Mouse, anti-Human, Clone: 2F4E10, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Spermidine synthase |
|---|---|
| Clone | 2F4E10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075145
|
Proteintech
68548-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18065395
|
Proteintech
68548-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Spermidine synthase antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Spermidine synthase. This antibody recognizes human antigen. The Spermidine synthase antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| Spermidine synthase | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| PAPT, SPDSY, spermidine synthase, SPS1, SRM, SRML1 | |
| Primary |
| 2F4E10 | |
| Western Blot, Immunofluorescence, ELISA | |
| Mouse | |
| 6723 | |
| IgG2b | |
| Store at -20°C. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.