missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINT1 Monoclonal Antibody (OTI1D2), TrueMAB™, OriGene
Description
SPINT1 Monoclonal antibody specifically detects SPINT1 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
| Antigen | SPINT1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | SCFD1 |
| Gene Accession No. | Q62991, Q8WVM8 |
| Gene Alias | HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?