missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRP54 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17171554 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17171554 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17171554 Supplier Invitrogen™ Supplier No. PA5113697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ENFEIIIVDT SGRHKQEDSL FEEMLQVANA IQPDNIVYVM DASIGQACEA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRP54
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SRP54
Gene Accession No. P61011
Gene Alias 54K subunit of signal recognition particle; 54kDa; signal recognition particle 54; signal recognition particle 54 kDa protein; signal recognition particle 54A; signal recognition particle 54kD; signal recognition particle 54kDa; SRP54; Srp54a
Gene Symbols SRP54
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SRP54.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6729
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.