Learn More
SSEA-5 Mouse, anti-Human, FITC, Clone: 8e11, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60063FI
Encompass_Preferred
 View more versions of this product
Product Code. 17188791
  • 653.40€ / 100 tests
Description
  • The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
  • Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
  • The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures
Specifications
Specifications
SSEA-5
Monoclonal
FITC
100 Tests
Human
Pluripotent Stem Cells
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
8e11
Mouse
Primary
IgG1 κ