missing translation for 'onlineSavingsMsg'
Learn More

SSEA-5 Mouse, anti-Human, FITC, Clone: 8e11, STEMCELL Technologies

Mouse Monoclonal Antibody

POA - 653.40€

Specifications

Antigen SSEA-5
Clone 8e11
Applications Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17198791
Encompass_Preferred
View Documents
Stemcell Technologies
60063FI.1
25 Tests This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
17188791
Encompass_Preferred
View Documents
Stemcell Technologies
60063FI
100 Tests
653.40€
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
  • Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
  • The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures
Specifications

Specifications

SSEA-5
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
FITC
Human
Primary
8e11
Monoclonal
Mouse
IgG1 κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.