Learn More
SSEA-5 Mouse, anti-Human, FITC, Clone: 8e11, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60063FI.1
Description
- The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells
- Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation
- The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures
Specifica
| SSEA-5 | |
| Monoclonal | |
| FITC | |
| 25 Tests | |
| Human | |
| Pluripotent Stem Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 8e11 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.