missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSR1 Monoclonal Antibody (OTI 5D5), TrueMAB™, OriGene
Description
SSR1 Monoclonal antibody specifically detects SSR1 in Canine, Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
| Antigen | SSR1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI 5D5 |
| Concentration | 0.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | SSR1 |
| Gene Accession No. | P16967, P43307 |
| Gene Alias | 2510001K09Rik; 6330400D04; Ac2-238; AI159733; AI452176; cb758; DKFZp781N23103; liver regeneration-related protein LRRG137; PGP35; PSEC0262; signal sequence receptor subunit 1; signal sequence receptor subunit alpha; signal sequence receptor, alpha; signal sequence receptor, alpha (translocon-associated protein alpha); SSR; SSR alpha subunit; SSR1; SSRA precursor protein; SSR-alpha; translocon associated protein alpha; translocon-associated protein alpha; translocon-associated protein alpha subunit; translocon-associated protein subunit alpha; TRAP alpha; TRAPA; TRAP-alpha; unnamed protein product |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?