missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSR2 Monoclonal Antibody (4C1)

Description
Peptide Sequence: SSAALDVELS DDSFPPEDFG IVSGMLNVKW DRIAPASNVS HTVVLRPLKA GYFNFTSATI TYLAQEDGPV VIGSTSAPGQ GGILAQREFD RRFSPHFLDW.
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2).The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein (alpha-SSR or SSR1) and a 22-kD glycoprotein (beta-SSR or SSR2). The human beta-signal sequence receptor gene (SSR2) maps to chromosome bands 1q21-q23.
Specifications
Specifications
| Antigen | SSR2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4C1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Ssr2 |
| Gene Accession No. | P43308 |
| Gene Alias | 1500032E05Rik; AI315033; AU020133; DKFZp686F19123; glycoprotein 25H; gp25H; HSD25; signal sequence receptor beta subunit; signal sequence receptor subunit 2; signal sequence receptor subunit beta; signal sequence receptor, beta; signal sequence receptor, beta (translocon-associated protein beta); signal sequence receptor, beta precursor; Ssr2; SSR2 protein; SSR-beta; TLAP; translocon-associated protein beta; translocon-associated protein subunit beta; translocon-associated protein subunit bet-like protein; TRAP[b]; TRAPb; TRAPbeta; TRAP-beta; wu:fb75b04; zgc:92180 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?