missing translation for 'onlineSavingsMsg'
Learn More

Standard Buffer Phosphate, pH 4.002, NIST, Chem-Lab

Product Code. 11908494
Change view
Click to view available options
Quantity:
1 L
10 L
Unit Size:
10 L
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11908494 10 L 10 L
11964004 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11908494 Supplier Chem-Lab Supplier No. CL03.0602.9010
  • ONLINE EXCLUSIVE PRICE
    332.00€
    171.00€ / 10 L
    Save 161.00€
    48% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For laboratory use, pH measurements, traceable to SRM from NIST

pH = 4.003 at 0°C
pH = 3.99 at 5°C
pH = 3.998 at 10°C
pH = 3.999 at 15°C
pH = 4.002 at 20°C
pH = 4.008 at 25°C
pH = 4.015 at 30°C
pH = 4.035 at 40°C
pH = 4.060 at 50°C
pH = 4.091 at 60°C
pH = 4.126 at 70°C
pH = 4.164 at 80°C

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Density 1.00g/mL
pH 4.002
Quantity 10 L
Packaging PE
Traceability to NIST NIST traceable
Formula Weight 204.23g/mol
Chemical Name or Material Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.