Learn More
DWK Life Sciences Stopcock Key, threaded, SBW-J glass, ground
Product Code. 15894518 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
Plug Bore:
2 mm
2.5 mm
4 mm
6 mm
Unit Size:
Pack of 25
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Plug Bore unitSize
15894518 14.5 NS 2.5 mm Pack of 50
15901511 12.5 NS 2.5 mm Pack of 50
15874518 12.5 NS 2 mm Pack of 50
15804528 14.5 NS 4 mm Pack of 50
15814528 18.8 NS 4 mm Pack of 25
15834528 18.8 NS 6 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15894518 Supplier DWK Life Sciences Supplier No. 286037505
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
The most common tap plug in the ground DURAN™ single way stopcock.
Solid SBW-J glass tap plugs have a thread on which the 3-part screw lock is mounted. The tap plugs are offered in 3 NS sizes and each NS size in two bore variants.
  • With thread, ground
  • Solid tap plug made of SBW-J glass
  • The tap plugs are offered in 3 NS sizes and each NS size in two bore variants
  • Assembly and thus securing of the valve plug by means of the 3-part screw locking device
  • Taper 1:10
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 2.5 mm
Material SBW-J Glass
Autoclavable Not Autoclavable
Closure Size 14.5 NS
No. per Pack 50/Pk.