missing translation for 'onlineSavingsMsg'
Learn More

Streptidine Sulfate Salt, TRC

Product Code. 30424253
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424253 100 mg 100mg
30417271 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424253 Supplier Toronto Research Chemicals Supplier No. TRCS687400100MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Streptidine Sulfate Salt
Molecular Formula C8H18N6O4 xH2SO4
Quantity 100 mg
Synonym N1,N3-Bis(aminoiminomethyl)streptamine Sulfate Salt, N,N'-Diamidinostreptamine Sulfate Salt, N,N'''-(2,4,5,6-Tetrahydroxy-1,3-cyclohexanediyl)bis-guanidine Sulfate Salt
SMILES O[C@H]1[C@H](O)[C@@H](NC(N)=N)[C@H](O)[C@@H](NC(N)=N)[C@@H]1O
Molecular Weight (g/mol) 262.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.