missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, TRC

Product Code. 30136332
Change view
Click to view available options
Quantity:
50 mg
5 g
10 g
Unit Size:
10g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136332 5 g 5g
30137097 10 g 10g
18092220 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136332 Supplier Toronto Research Chemicals Supplier No. TRCS6990845G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-79-1
Molecular Formula C18 H14 N4 O5 S
Molecular Weight (g/mol) 398.39
Synonym Sulfasalazine, 2-Hydroxy-5-[2-[4-(pyridin-2-ylsulfamoyl)phenyl]diazenyl]benzoic acid, Benzoic acid, 2-hydroxy-5-[2-[4-[(2-pyridinylamino)sulfonyl]phenyl]diazenyl]-Benzoic acid, 2-hydroxy-5-[[4-[(2-pyridinylamino)sulfonyl]phenyl]azo]- (9CI), Salicylic acid, 5-[[p-(2-pyridylsulfamoyl)phenyl]azo]- (6CI,7CI,8CI), 2-Hydroxy-5-((4-((2-pyridinylamino)sulfonyl)phenyl)azo)benzoic acid, 5-[4-(2-Pyridylsulfamyl)phenylazo]-2-hydroxybenzoic acid, 5-[p-(2-Pyridylsulfamoyl)phenylazo]salicylic acid, 5-[p-(2-Pyridylsulfamyl)phenylazo]salicylic acid, Azopyrin, Azopyrine, Azulfidine, Azulfidine EN, Azulfin, Benzosulfa, Colo-Pleon, NSC 203730, NSC 667219, Reupirin, Salazopyridin, Salazopyrin, Salazopyrine, Salazosulfapyridin, Salazosulfapyridine, Salicylazosulfapyridine, Salisulf, Sulfasalazin, Sulfasalazine, Sulphasalazine
IUPAC Name 2-hydroxy-5-[(E)-[4-(pyridin-2-ylsulfamoyl)phenyl]diazenyl]benzoic acid
SMILES OC(=O)c1cc(ccc1O)N=Nc2ccc(cc2)S(=O)(=O)Nc3ccccn3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H14N4O5S/c23-16-9-6-13(11-15(16)18(24)25)21-20-12-4-7-14(8-5-12)28(26,27)22-17-3-1-2-10-19-17/h1-11,23H,(H,19,22)(H,24,25)/b21-20+
Formula Weight 398.0685
Chemical Name or Material Sulfasalazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.