missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 18.0 N, Ricca Chemical
Specifications
Specifications
| Appearance of Solution | Colorless Liquid |
| CAS Min % | 51.0, 47.0 |
| Color | Colorless |
| CAS Max % | 53.0, 48.92 |
| Concentration | 17.95 to 18.05 N |
| Packaging | Natural Poly Bottle |
| Quantity | 4 L |
| Physical Form | Liquid |
| Synonym | oil of vitriol |
| Grade | Laboratory |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?