missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+ atom % D

Product Code. 10205660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205660 100 mL Glass Bottle 100mL
10387830 50 mL Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10205660 Supplier Thermo Scientific Acros Supplier No. 175481000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 13813-19-9 | H2O4S | 100.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 13813-19-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 100.08
MDL Number MFCD00003542
InChI Key QAOWNCQODCNURD-ZSJDYOACSA-N
Synonym sulfuric acid-d2, deuterosulfuric acid, dideuterosulfuric acid, sulphuric 2h2 acid, sulfuric acid-d2 solution, d2so4, dideuterosulfuric acid solution, 2 h? sulfuric acid, sulfuric acid-d2, 98 wt% in deuterium oxide, deuterosulfuric acid min in d 2 o isotopic
PubChem CID 160992
IUPAC Name dideuterio sulfate
SMILES [2H]OS(=O)(=O)O[2H]

Spezifikation

Physical Form Liquid
Color Colorless
Density 1.8700g/mL
Deuterium (D) 99.50% min.
Packaging Glass Bottle
Quantity 100 mL
Linear Formula D2SO4
Formula Weight 100.09
Percent Purity 97.5 to 102.5%
Grade NMR
Chemical Name or Material Sulfuric acid-d2, 99.5+ atom % D
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Sulfuric acid-d2, 98 wt.% in D2O, 99.5+ atom % D
Signalwort
  • Danger
Gefahrenkategorie
  • Skin corrosion/irritation Category 1 A
Gefahrenhinweis
  • H314-Causes severe skin burns and eye damage.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.