Description
This membrane body is a replacement part for use with InPro™ O2 sensors. The InPro family provides the means for reliable inline measurement of dissolved oxygen in a wide spectrum of processes that operate under sterile/hygienic conditions. To meet the range of respective requirements, a selection of advanced O2 sensors and measuring systems is available.
- The InPro 6800 is used in many applications in brewing and the beverage industry
- The InPro 6050 is often used in water/wastewater treatment and fish farming
- Replacement membrane bodies are supplied in packs of 16
- All membrane bodies are supplied complete with material certificate
Specifications
Specifications
| Description | Pack of 16 |
| For Use With (Equipment) | InPro™ O2 Sensors |
| Type | Oxygen Membrane Body |
| Material | PTFE-coated Stainless Steel |