missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAFA3/FAM19A3 Antibody (460904) [Janelia Fluor™ 525], Novus Biologicals™
Shop All Bio Techne ProductsDescription
TAFA3/FAM19A3 Monoclonal antibody specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
Specifications
| Antigen | TAFA3/FAM19A3 |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), CyTOF |
| Classification | Monoclonal |
| Clone | 460904 |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Chemokine-like protein TAFA-3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human TAFA3/FAM19A3, Ala31-Arg133, Accession # Q7Z5A8 |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?