missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBCE Polyclonal Antibody

Description
Immunogen sequence: LEKLPSLRAL SCLRNPLTKE DKEAETARLL IIASIGQLKT LNKCEILPEE RRRAELDYRK AFGNEWKQAG GHKDPEKNRL SEEFLTAHPR YQFLCLKYGA PEDWELKTQQ PLMLKNQLLT LKIKYPHQLD QKVLEKQLPG SMTIQKVKGL LSRLLKVPVS DLLLSYESPK KPGREIELEN DLKSLQFYSV ENGDCLLVRW.
Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Specifications
Specifications
| Antigen | TBCE |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 5.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TBCE |
| Gene Accession No. | Q15813 |
| Gene Alias | 2610206D02Rik; beta-tubulin cofactor E; C530005D02Rik; HRD; hypoparathyroidism, growth and mental retardation, and dysmorphism; hypothetical protein LOC664760; KCS; KCS1; pac2; pmn; Tbce; Tbce protein; tubulin folding cofactor E; tubulin-folding cofactor E; Tubulin-specific chaperone E; tubulin-specific chaperone E-like protein; Unknown (protein for MGC:133938); zgc:123075 |
| Gene Symbols | TBCE |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?