missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-2/HNF-1 beta Antibody (CL0374), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-30678-25ul
This item is not returnable.
View return policy
Description
TCF-2/HNF-1 beta Monoclonal specifically detects TCF-2/HNF-1 beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TCF-2/HNF-1 beta | |
| Monoclonal | |
| Unconjugated | |
| P35680 | |
| HNF1B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KEVLVQALEELLPSPNFGVKLETLPLSPGSGAEPDTKPVFHTLTNGHAKGRLSGDEGSEDGDDYDTPPILKELQALNTEEAAEQRAEVDRMLSEDPWRAAKMIKGYMQQH | |
| 25 μL | |
| Diabetes Research | |
| 6928 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL0374 | |
| Western Blot 1 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 ug/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| FJHN, hepatocyte nuclear factor 1-beta, HNF1 beta A, HNF1 homeobox B, HNF-1B, HNF1beta, HNF-1-beta, HNF2, Homeoprotein LFB3, LFB3, LF-B3, MODY5HPC11, TCF-2, TCF2transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor, Transcription factor 2, transcription factor 2, hepatic, Variant hepatic nuclear factor 1, vHNF1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur