missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-2/HNF-1 beta Antibody (CL0374), Novus Biologicals™
Mouse Monoclonal Antibody
369.00€ - 554.00€
Specifications
| Antigen | TCF-2/HNF-1 beta |
|---|---|
| Clone | CL0374 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18439131
|
Novus Biologicals
NBP2-30678-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18708233
|
Novus Biologicals
NBP2-30678 |
0.1 mL |
554.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCF-2/HNF-1 beta Monoclonal specifically detects TCF-2/HNF-1 beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TCF-2/HNF-1 beta | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Diabetes Research | |
| P35680 | |
| 6928 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KEVLVQALEELLPSPNFGVKLETLPLSPGSGAEPDTKPVFHTLTNGHAKGRLSGDEGSEDGDDYDTPPILKELQALNTEEAAEQRAEVDRMLSEDPWRAAKMIKGYMQQH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL0374 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FJHN, hepatocyte nuclear factor 1-beta, HNF1 beta A, HNF1 homeobox B, HNF-1B, HNF1beta, HNF-1-beta, HNF2, Homeoprotein LFB3, LFB3, LF-B3, MODY5HPC11, TCF-2, TCF2transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor, Transcription factor 2, transcription factor 2, hepatic, Variant hepatic nuclear factor 1, vHNF1 | |
| HNF1B | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title