Learn More
TDP-43 (C-terminal) Rabbit, anti-Human, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech BIOTIN-12892-20UL
 View more versions of this product
Product Code. 18069795
  • 145.00€ / 20µL
Description
The TDP-43 (C-terminal) antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TDP-43 (C-terminal). This antibody recognizes human, mouse, rat antigen. The TDP-43 (C-terminal) antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specifications
Specifications
TDP-43 (C-terminal)
1000 μg/mL
ALS10, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43
20 μL
23435
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunohistochemistry
Biotin
Rabbit
Primary
Human, Mouse, Rat
150 kDa