Learn More
TDP-43 (C-terminal) Rabbit, anti-Human, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech BIOTIN-12892-20UL
Description
The TDP-43 (C-terminal) antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TDP-43 (C-terminal). This antibody recognizes human, mouse, rat antigen. The TDP-43 (C-terminal) antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specifications
| TDP-43 (C-terminal) | |
| 1000 μg/mL | |
| ALS10, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43 | |
| 20 μL | |
| 23435 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunohistochemistry | |
| Biotin | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |