Learn More
TDP-43 (C-terminal) Rabbit, anti-Human, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | TDP-43 (C-terminal) |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry |
| Conjugate | Biotin |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18086675
|
Proteintech
BIOTIN-12892-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18069795
|
Proteintech
BIOTIN-12892-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TDP-43 (C-terminal) antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TDP-43 (C-terminal). This antibody recognizes human, mouse, rat antigen. The TDP-43 (C-terminal) antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specifications
| TDP-43 (C-terminal) | |
| Immunohistochemistry | |
| Liquid | |
| Human, Mouse, Rat | |
| ALS10, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43 | |
| Primary |
| 1000 μg/mL | |
| Biotin | |
| Rabbit | |
| 23435 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.