missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TEF5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15544812
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15544812 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15544812 Supplier Invitrogen™ Supplier No. PA541447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: MMNSVLENFT ILQVVTSRDS QETLLVIAFV FEVSTSEHGA QHHVYKLVKD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is predominantly expressed in the placenta and is involved in the transactivation of the chorionic somatomammotropin-B gene enhancer. Translation of this protein is initiated at a non-AUG (AUA) start codon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEF5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TEAD3
Gene Accession No. Q99594
Gene Alias DTEF-1; ETFR-1; ETF-related factor 1; Tcf13r2; TEA domain family member 3; TEA domain family member 5; TEA domain transcription factor 3; Tead3; TEAD-3; TEAD5; TEF5; TEF-5; transcriptional enhancer factor 5; transcriptional enhancer factor TEF-5; transcriptional enhancer factor TEF-5 (DTEF-1)
Gene Symbols TEAD3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TEAD3 (aa 386-435).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7005
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.