missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl propargyl ether, 98%

Product Code. 15420348
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420348 100 g 100g
15400348 5 g 5g
15410348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420348 Supplier Thermo Scientific Alfa Aesar Supplier No. H53480.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16314-18-4 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16314-18-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD07368651
InChI Key QIGSPXPODKLGRK-UHFFFAOYSA-N
PubChem CID 7172302
IUPAC Name 2-methyl-2-prop-2-ynoxypropane
SMILES CC(C)(C)OCC#C

Specifications

Density 0.835
Boiling Point 119°C
Flash Point 20°C (68°F)
Linear Formula HC≡CCH2OC(CH3)3
Refractive Index 1.418
Quantity 100 g
UN Number UN1993
Synonym tert-butyl propargyl ether, 3-tert-butoxy prop-1-yne, 1-propyne,3-1,1-dimethylethoxy, 1-propyne, 3-1,1-dimethylethoxy, 3-tert-butoxyprop-1-yne, 3-tert-butyloxy-1-propyne, tert-butylpropargylether
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material tert-Butyl propargyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.