missing translation for 'onlineSavingsMsg'
Learn More

Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water

Product Code. 11454127
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454127 1000 g 1000g
11483930 100 g 100g
11464127 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454127 Supplier Thermo Scientific Alfa Aesar Supplier No. A19560.0B
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 67-03-8 | C12H18Cl2N4OS | 337.263 g/mol

Thiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.

Solubility
Soluble in water, glycerol, ethanol, dimethyl sulfoxide and methanol. Insoluble in benzene, hexane, chloroform and ether.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Molecular Weight (g/mol) 337.263
MDL Number MFCD00012780
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
PubChem CID 6202
ChEBI CHEBI:49105
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]

Specifications

Melting Point ∼250°C (decomposition)
Assay Percent Range 0.99
Quantity 1000 g
Beilstein 3851771
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9295
Solubility Information Soluble in water,glycerol,ethanol,dimethyl sulfoxide and methanol. Insoluble in benzene,hexane,chloroform and ether.
Formula Weight 337.28
Percent Purity 99%
Assay (dry wt.), may cont. up to 5% water
Chemical Name or Material Thiamine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.