missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-3-carboxaldehyde, 96%

Product Code. 11487576
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487576 10 g 10g
11497576 50 g 50g
11407586 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487576 Supplier Thermo Scientific Alfa Aesar Supplier No. A14628.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 498-62-4 | C5H4OS | 112.146 g/mol

Thiophene-3-carboxaldehyde has been used in the synthesis of series of 4-substituted 2-thiophenesulfonamides, acetal and ketal derivatives of 4?-demethylepipodophyllotoxin-β-D-glucoside and epipodophyllotoxin-β-D-glucoside. Also has been used to produce 1,2-di-3-thienyl-2-hydroxyethanone (3,3?-thenoin), 3-thienyl symmetric analog of benzoin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-3-carboxaldehyde has been used in the synthesis of series of 4-substituted 2-thiophenesulfonamides, acetal and ketal derivatives of 4′-demethylepipodophyllotoxin-β-D-glucoside and epipodophyllotoxin-β-D-glucoside. Also has been used to produce 1,2-di-3-thienyl-2-hydroxyethanone (3,3′-thenoin), 3-thienyl symmetric analog of benzoin.

Solubility
Not miscible in water.

Notes
It is sensitive to air. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-62-4
Molecular Formula C5H4OS
Molecular Weight (g/mol) 112.146
MDL Number MFCD00005466
InChI Key RBIGKSZIQCTIJF-UHFFFAOYSA-N
Synonym 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde
PubChem CID 68135
ChEBI CHEBI:87611
IUPAC Name thiophene-3-carbaldehyde
SMILES C1=CSC=C1C=O

Specifications

Density 1.28
Boiling Point 194°C to 196°C
Flash Point 64°C (147°F)
Refractive Index 1.584
Quantity 10 g
Beilstein 105889
Sensitivity Air sensitive
Solubility Information Not miscible in water.
Formula Weight 112.15
Percent Purity 96%
Chemical Name or Material Thiophene-3-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • Thiophene-3-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Thiophenecarboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.