missing translation for 'onlineSavingsMsg'
Learn More

EnquireBio TIGIT Mouse, APC, Clone: 1G9, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB57-APC-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932228

  • 128.00€ / 25µg

Estimated Shipment: 08-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

TIGIT
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Tigit
Protein A/G Purified
RUO
100043314
IgG1 κ
Flow Cytometry
1G9
APC
XP_017172323
Mouse
25 μg
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions