missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TIGIT Recombinant Llama Monoclonal Antibody (2C7)
GREENER_CHOICE

Product Code. 17931811
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931811 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17931811 Supplier Invitrogen™ Supplier No. MA549357

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Llama Recombinant Monoclonal Antibody

TIGIT Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow, ELISA

TIGIT is a 26 kDa protein that belongs to the poliovirus receptor (PVR) family. It is expressed on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but not on B cells or naive CD4+ T cells. However, TIGIT can be upregulated on naive CD4+ T cells upon activation. TIGIT interacts with members of the PVR and PVR-like families, including CD155. This interaction mediates the engagement of NK and T cells with antigen-presenting cells, fibroblasts, and endothelial cells expressing PVR and PVR-like proteins. Binding of TIGIT to PVR leads to increased secretion of IL-10 and decreased secretion of IL-12B, promoting the generation of mature immunoregulatory dendritic cells and suppressing T cell activation. Through these interactions, TIGIT plays a significant role in modulating immune responses and maintaining immune homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIGIT
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 2C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TIGIT
Gene Accession No. Q495A1
Gene Alias ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM
Gene Symbols TIGIT
Host Species Llama
Immunogen A recombinant protein corresponding to amino acids 22-141 of human TIGIT.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201633
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype scFv
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.