Learn More
TIMP-2 Antibody (D08-1D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11415-0.1mg
 View more versions of this product
Product Code. 18612647
  • 479.00€ / 0.10mg
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TIMP-2
Monoclonal
Unconjugated
CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2
E. coli derived recombinant protein corresponding to aa 27-220 of human TIMP-2
0.1 mg
Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers
7077
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
ELISA
D08-1D4
PBS
Mouse
Protein G purified