missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Antibody (D08-1D4), Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 479.00€
Specifications
| Antigen | TIMP-2 |
|---|---|
| Clone | D08-1D4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663788
|
Novus Biologicals
NBP3-11415-0.025mg |
0.025 mg |
203.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612647
|
Novus Biologicals
NBP3-11415-0.1mg |
0.1 mg |
479.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human samples. It is validated for ELISASpecifications
| TIMP-2 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| PBS | |
| 7077 | |
| IgG1 | |
| Protein G purified |
| D08-1D4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| E. coli derived recombinant protein corresponding to aa 27-220 of human TIMP-2 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title