missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Antibody (D08-1D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11415-0.025mg
This item is not returnable.
View return policy
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Human samples. It is validated for ELISA
Specifications
| TIMP-2 | |
| Monoclonal | |
| Unconjugated | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| E. coli derived recombinant protein corresponding to aa 27-220 of human TIMP-2 | |
| 0.025 mg | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 7077 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| D08-1D4 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction