Learn More
Tin(II) methanesulfonate, 50% w/w aq. soln.
Product Code. 15466178
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466178 100 mL 100mL
15476178 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15466178 Supplier Thermo Scientific Alfa Aesar Supplier No. H59773.AE
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
CAS: 53408-94-9 | C2H6O6S2Sn | 308.89 g/mol
It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).
Solubility
Not miscible or difficult to mix in water.
Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53408-94-9
Molecular Formula C2H6O6S2Sn
Molecular Weight (g/mol) 308.89
MDL Number MFCD00137737
InChI Key JALQQBGHJJURDQ-UHFFFAOYSA-L
Synonym stannous methanesulfonate, methanesulfonic acid, tin 2+ salt, tin ii methanesulfonate, methanesulfonate; tin 2+, methanesulfonic acid, tin 2+ salt 2:1, tin ii methanesulfonate solution, tin ii bis methanesulfonate, pubchem11154, stannous methylsulfonate, 2ch3o3s.sn
PubChem CID 6452824
IUPAC Name methanesulfonate;tin(2+)
SMILES [Sn++].CS([O-])(=O)=O.CS([O-])(=O)=O
Specifications
Melting Point -27°C
Density 1.55
Linear Formula (CH3SO3)2Sn
Refractive Index 1.444
Quantity 100 mL
UN Number UN3265
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 308.91
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material Tin(II) methanesulfonate